This item is now discontinued. View the direct replacement: EW-20020-96 or browse similar items
Standard Sensitivity Noncontact Probe
Representative image only.
Item # EW-20020-95
Fast troubleshooting during equipment operation (safety permitting)
  • Isolates problems as electrical or mechanical in seconds without a schematic
  • Requires no direct electrical connection
  • Variable sensitivity
  • Detects troublesome transient pulses
  • Every unit hand-crafted and quality control-tested
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStandard Sensitivity Noncontact Probe
  • Warranty1 Month
Key Features
  • Isolates problems as electrical or mechanical in seconds without a schematic
  • Requires no direct electrical connection
  • Variable sensitivity
  • Detects troublesome transient pulses
  • Every unit hand-crafted and quality control-tested
  • Can be used in high-risk explosive environments
  • Long-life power source and low current LED lasts for at least 5000, 5 second operations
  • Detects residual magnetism
  • Shelf life of 5 years
  • 1 month warranty
More About this Item
Mag-Probe® is a magnetic sensing instrument used to trouble-shoot AC and DC powered solenoids, solenoid valves, relays, mercury wetted relays, heat controllers and test coils used in robotics— even automotive alternators, voltage regulators and electronic ignitions systems. Unit is ideal for testing the largest to sub-miniature solenoids, relays and most reed relays and all automotive applications.