Starrett 3600-5 Basic Electroinic Indicator, 0.5in (Depth Gauges/Indicators)

Representative image only.
Starrett Mfr # 3600-5 – Item # EW-09925-33
  • Range: 0.500"/12.7 mm
  • Resolution: 0.005”/0.01 mm
  • AGD group 2 size
  • Reverse travel
  • Zero setting at any position

$234.00USD / Each

$82.00USD / Each

for an additional
$82.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionBasic Electroinic Indicator, 0.5in (Depth Gauges/Indicators)

Key Features

  • Range: 0.500"/12.7 mm
  • Resolution: 0.005”/0.01 mm
  • AGD group 2 size
  • Reverse travel
  • Zero setting at any position
  • Lug-on-center back and additional flat back
  • Includes dust cap

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat