Starrett 98-6 6" Machinists' Level with ground and graduated vials
Representative image only.
Starrett Mfr # 98-6 – Item # EW-09925-55
Adjustable vials offer positive setting
  • Six inch (150 mm) rail length
  • Base is precision-machined seasoned cast iron
  • Vials are adjustable to a positive setting
  • Housed in satin finished brass tube with friction-fit closing cover to prevent breakage
  • 1 year warranty
$177.00USD / Each
$99.00 - $175.50USD / Each
for an additional
$99.00USD / Each
for an additional
$175.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Description6" Machinists' Level with ground and graduated vials
  • Warranty1 year
1 Year Warranty
Key Features
  • Six inch (150 mm) rail length
  • Base is precision-machined seasoned cast iron
  • Vials are adjustable to a positive setting
  • Housed in satin finished brass tube with friction-fit closing cover to prevent breakage
  • 1 year warranty
More About this Item
Involute groove running the length of the level base provides reliable seating in ground work such as pipes
or shafting. Cross test vials allow simultaneous leveling in both directions.