Starrett S766AZ Electronic 6" Caliper and 1" Micrometer Set
Representative image only.
Starrett Mfr # S766AZ – Item # EW-09925-29
  • 1 year warranty
$505.00USD / Each
$45.50 - $121.50USD / Each
for an additional
$45.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectronic 6" Caliper and 1" Micrometer Set
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Electronic Caliper & Micrometer Measuring Set
6" Electronic caliper — 0.0005"/0.01 mm resolution
0 to 1"/25 mm Electronic micrometer — 0.0005"/0.001 mm through 4", 0.001"/0.001 mm above 4"