Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/box

Representative image only.
Item # EW-06139-00

$171.00USD / of 500

BUY MORE & SAVE

  • 1-5$171.00USD / of 500
  • 6-29$153.90USD / of 500
  • 30-119$145.35USD / of 500
  • 120+$136.80USD / of 500

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolystyrene
  • Overall Height (in)9/16
  • Overall Height (cm)1.5
  • Outside Diameter (OD) (in)3 15/16
  • Outside Diameter (OD) (cm)10
  • DescriptionSterile Petri Dishes; 100 mm dia x 15 mm H; 500/box

More About this Item

Disposable polystyrene dishes have convenient stacking ridges. Packed sterile in easy-open bags of 20. Gamma irradiated. 2 bx/cs*.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat