Scienceware Stir Bar Retriever, Flexible PTFE; 14-3/4" Long
Representative image only.
Scienceware Mfr # 37774-0101 – Item # EW-08555-20
Eliminates messy spills and breakage
  • Flexible shaft makes retrieving those hard-to-reach stir bars easy
  • 2 year warranty
PRODUCT OPTIONS
$43.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)14 3/4
  • DescriptionStir Bar Retriever, Flexible PTFE; 14-3/4\" Long
Two year warranty
Key Features
  • Flexible shaft makes retrieving those hard-to-reach stir bars easy
  • 2 year warranty
More About this Item
Easily retrieve stir bars without spills or possible breakage. The alnico V magnet is completely enclosed.