Stir-Pak Analog Speed Controller; 115 VAC
Representative image only.
Stir-Pak Item # EW-50007-00
Easily customize to your needs
  • Simple controls—speed adjustment knob and separate "forward/off/reverse" switch
  • Reduce gear failure with torque limiting protection
$408.00USD / Each
$150.00USD / Each
for an additional
$150.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
Two year warranty
Key Features
  • Simple controls—speed adjustment knob and separate "forward/off/reverse" switch
  • Reduce gear failure with torque limiting protection
More About this Item
Controllers: Solid-state controller offers precise speed control with a 100:1 adjustment range. A separate ''forward/off/reverse'' switch works independently from the speed control dial—allowing you to repeat mixer speed settings from one run to the next. Electronic control maintains speed regulation better than ±2%. Torque limiting provides greater system protection and reduced motor gear failure. An LED indicates when power is on. Controller housing is IP23 rated.
Remote controller is connected to the mixer head via a 6-ft (1.8-m), three-wire cord—protects electronics from hazardous spills or splashes. Controller measures 7'' W x 3 3/16'' H x 4 7/8'' D (178 mm W x 81 mm H x 124 mm D).
What's included: 115 VAC model has a 6-ft (1.8-m) cord with grounded plug. The 230 VAC model has an 8-ft (2.4-m) cord with modular line cord and IEC320/CEE22 connector. When ordering, specify country of destination to receive correct line cord.