Stopcock with Compression Fittings, 1/4" tube OD
Representative image only.
Item # EW-06373-86
Get a secure seal at higher pressures
  • Constructed of chemically inert PTFE and PFA
PRODUCT OPTIONS
$140.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" compression
  • Connection TypeCompression
  • Inlet Port1/4" compression
  • Cv0.44
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • Tubing OD (in)0.25
  • DescriptionStopcock with Compression Fittings, 1/4" tube OD
Key Features
  • Constructed of chemically inert PTFE and PFA
More About this Item
Stopcocks have body made of PFA and plug made of PTFE. Maximum pressure is 50 psi.