Stopcock with Compression Fittings, 1/8" tube OD
Representative image only.
Item # EW-06373-85
Get a secure seal at higher pressures
  • Constructed of chemically inert PTFE and PFA
PRODUCT OPTIONS
$143.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/8" compression
  • Connection TypeCompression
  • Inlet Port1/8" compression
  • Cv0.38
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • Tubing OD (in)0.125
  • DescriptionStopcock with Compression Fittings, 1/8" tube OD
Key Features
  • Constructed of chemically inert PTFE and PFA
More About this Item
Stopcocks have body made of PFA and plug made of PTFE. Maximum pressure is 50 psi.