Straight threaded coupling, PFA, 1/4" NPT(F)
Representative image only.
Item # EW-06546-01
Create a secure pipe threaded connection
  • Constructed of chemically inert PFA
PRODUCT OPTIONS
$51.00USD / Each
BUY MORE & SAVE
  • 1-9$51.00USD / Each
  • 10-49$45.90USD / Each
  • 50-99$43.35USD / Each
  • 100+$40.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodEthylene oxide (ETO) only
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)1/4
  • Thread size NPT(F) (port 2) (in)1/4
  • DescriptionStraight threaded coupling, PFA, 1/4" NPT(F)
Key Features
  • Constructed of chemically inert PFA
More About this Item
These straight threaded coupling fittings are made of chemically inert PFA. Fittings can be sterilized via autoclave or ethylene oxide (ETO).