Stuart Hybridization Bottle, 6.8" x 1.5"

Representative image only.
Stuart Item # EW-39454-99
A wide range of options for any application

PRODUCT OPTIONS

$108.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialGlass, polyester, PTFE
  • Compatible WithRotisserie: 39454-95
  • Length (in)6.8
  • Diameter (in)1.5
  • DescriptionHybridization Bottle, 6.8" x 1.5"

More About this Item

Choose from a wide range of accessories for your Stuart hybridization incubator. Choose from differnt capacity rotisseries and bottles to suit your specific applicaiton.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat