Cole-Parmer Stack Blocks for Roller Shakers; 4/PK
Representative image only.
Cole-Parmer Item # EW-04653-09
Stack up to 3 shakers for bench saving space!
  • Previous part number SRT/STACK
$467.25USD / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer RS-200 Roller Shakers
  • DescriptionStack Blocks for Roller Shakers, set of 4.
Key Features
  • Previous part number SRT/STACK
More About this Item
Kit contains four blocks. A single kit will stack two units — order two kits to stack three tube roller shakers.