Cole-Parmer Tube Rotator Test Tube Holder Disc, 20 Tube Capacity
Representative image only.
Cole-Parmer Item # EW-00077-15
Quickly and easily replace tube holder without tools
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers an end over end action
  • Holds 20 x 9 to 20 mm tubes
  • 1 year warranty
  • Previous part number SB3/2
PRODUCT OPTIONS
$241.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Tubes20
  • Tube Size9 to 20 mm dia
  • DescriptionTube Rotator Test Tube Holder Disc, 20 tube capacity
  • Warranty1 Year
1 Year Warranty
Key Features
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers an end over end action
  • Holds 20 x 9 to 20 mm tubes
  • 1 year warranty
  • Previous part number SB3/2