Suction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
Representative image only.
Item # EW-74604-42
Protect pump from sediments filtering into suction line
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
PRODUCT OPTIONS
$18.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPVC and Stainless Steel
  • DescriptionSuction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
Key Features
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
More About this Item
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.