Supco 6" Circular Chart Recorder Paper, 6 Hour, -40 to 0°C, for Type J Thermocouple Recorders; 60/Pack
Representative image only.
Supco Mfr # CR87-13 – Item # EW-80050-78
For Supco brand chart recorders
  • Circular and strip charts
  • For CR87BC and CR87JC series recorders
  • 6 month warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed6 hour
  • Chart Diameter (in)6
  • Max Temperature (° F)32
  • Min Temperature (° F)-40
  • Max Temperature (° C)0
  • Min Temperature (° C)-40
  • Description6" Circular Chart Recorder Paper, 6 Hour, -40 to 0°C, for Type J Thermocouple Recorders; 60/Pack
  • Warranty6 month
6 Month Warranty
Key Features
  • Circular and strip charts
  • For CR87BC and CR87JC series recorders
  • 6 month warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.