Supco E3050C12 6" Chart Paper, -30 to 50 C, 12 Hour; 60/Pk
Representative image only.
Supco Mfr # E3050C12 – Item # EW-80055-40
For Supco brand chart recorders
  • Circular and strip charts
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed12 hour
  • Chart Diameter (in)6
  • Max Temperature (° F)122
  • Min Temperature (° F)-22
  • Max Temperature (° C)50
  • Min Temperature (° C)-30
  • Description6\" Chart Paper, -30 to 50 C, 12 Hour; 60/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Circular and strip charts
  • 1 year warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.