Supco RPT61 Pressure/Temperature Chart Master
Representative image only.
Supco Mfr # RPT61 – Item # EW-18001-89
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPressure/Temperature Chart Master
More About this Item
For instant information, the PT Chart Master eliminates the need for traditional pressure / temperature charts. Accurate and easy to use, the RPT61 provides pressure and temperature correlation for 61 different refrigerants.
  • Choose pressure in PSI, BAR or Mpa
  • Choose temperature in F or C
  • 61 refrigerant tables
  • Large LCD
  • Auto off (after 5 minutes
  • Calculate superheat or sub-cooling