Supco SL500XT External Probe Temperature Data Logger with Real Time LCD
Representative image only.
Supco Mfr # SL500XT – Item # EW-18000-54
USB interface for fast data transfer
  • Real-time display of temperature, relative humidity (SL500TH only), time, date, alarm status, logging status, and battery life
  • Nonvolatile memory retains collected data—even if the battery fails
  • 1 year warranty
PRODUCT OPTIONS
$105.80USD / Each
$132.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature (one external)
  • Max Temperature (° F)176
  • Min Temperature (° F)-40
  • Max Temperature (° C)80
  • Min Temperature (° C)-40
  • Temperature Accuracy (° F)±0.9
  • Temperature Accuracy (° C)±0.5
  • Memory Size43344 datapoints
  • Battery Life3 years average use
  • Battery Type1 3V CR2032 lithium (included)
  • Display TypeLCD
  • DescriptionExternal Probe Temperature Data Logger with Real Time LCD
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Real-time display of temperature, relative humidity (SL500TH only), time, date, alarm status, logging status, and battery life
  • Nonvolatile memory retains collected data—even if the battery fails
  • 1 year warranty
More About this Item
Supco SL500 data loggers are compact, light weight, and easy to use—making them perfect for many applications. Each logger includes software needed for logger set up, data downloading, and analysis. Loggers can be programmed to record over points once the memory is full, or stop recording. Sampling rates can be adjusted from every 1 second to 9 hours, and start/stop may be manually controlled on the logger or a specific run time may be set through the software. The temperature and relative humidity model (18000-53) alternates display of temperature and humidity every two seconds.