Swift Optical M10DB-MP Phase Contrast Microscope with Digital Camera, Binocular, 115 VAC, 60 Hz

Representative image only.
Swift Optical Mfr # M10DB-MP – Item # EW-48925-04
Built-in, 3-megapixel camera captures still images or video onto your computer

$3,500.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Magnification100x/400x/1000x
  • Maximum Magnification1000x
  • Head ConfigurationBinocular
  • Resolution3.0 MP
  • Eyepiece Diameter (mm)20
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Stage Dimensions140 x 140 mm
  • Illuminator(s)LED, 3 W
  • Cable(s)USB
  • Power (VAC)115
  • DescriptionPhase Contrast Microscope with Digital Camera, Binocular, 115 VAC, 60 Hz

Includes

software, USB cable, and dust cover.

More About this Item

Use this phase contrast microscope to examine the fine details of transparent objects that possess very little contrast. You can also study unstained living organisms and cells without the use of fixing and staining reagents that destroy specimen or change their composition. Capture images using the built-in camera and upload to your computer through the USB output. Edit, measure, and annotate saved images with the included software.
Adjustable LED illumination provides bright, energy-efficient light; the multi-phase condenser with rack and pinion focusing enhances illumination for phase contrast viewing. Siedentopf binocular head allows easy adjustability of interpupillary distance while the 20-mm eyepieces give an increased field of view. The left eyepiece features diopter adjustment to compensate for the difference between a user's eyes. Built-in low-drive, mechanical stage features smooth-moving coaxial controls. All metal internal parts ensure long-term durability.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat