Talboys Sleeve for extension rods 04367-10, 04553-50 and -55
Representative image only.
Talboys Mfr # 171 – Item # EW-04364-00
$82.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Shaft MaterialStainless Steel
  • Shaft Diameter (in)0.3125
  • Shaft Diameter (mm)8
  • Shaft Length (in)3
  • Shaft Length (mm)76
  • DescriptionSleeve for extension rods 04367-10, 04553-50 and -55
More About this Item
Extension rods and sleeves increase the length of your shaft. One sleeve is required to connect each extension rod to your shaft.
Sleeve measures 3"L.