Target C-4000-1W DP Vials, clear with write-on patch

Representative image only.
Target Mfr # C4000-1W – Item # EW-98810-01
40% larger opening for problem-free sample insertion and extraction
  • A variety of vial styles and septa materials to match your equipment and application
  • Vials are 2-mL, 12 x 32 mm size

PRODUCT OPTIONS

$34.50USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)2
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeScrew-Thread
  • ThreadExternal
  • Height (in)1 17/64
  • Outside Diameter (in)15/32
  • Height (mm)32
  • Outside Diameter (mm)12
  • DescriptionDP Vials, clear with write-on patch

Key Features

  • A variety of vial styles and septa materials to match your equipment and application
  • Vials are 2-mL, 12 x 32 mm size

More About this Item

Get the assurance of a crimp seal with the time-saving convenience of a screw cap. The unique threaded crown design of these glass vials ensures a secure seal with just a half-turn of the cap. Precision formed neck and crimp cap profile are compatible with robotic arm autosampler trays.
Vials with write-on patches have graduations at 0.5, 1.0, and 1.5 mL. Select the septum appropriate for your application. Pre-slit septa are available for applications using a very thin (32-gauge) needle, or where a vacuum may form in the vial.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat