This item is now discontinued. We recommend the following as suggested replacements: EW-98810-00
Target Screw-Thread Vials, clear with write-on patch
Representative image only.
Target Mfr # C4010-1W – Item # EW-98813-01
Threaded vials offer a secure closure
  • Manufactured from clear, Type 1 Class A glass and a 10-425 thread finish
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)2
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeScrew-Thread
  • Height (in)1 17/64
  • Height (mm)32
  • DescriptionScrew-Thread Vials, clear with write-on patch
Key Features
  • Manufactured from clear, Type 1 Class A glass and a 10-425 thread finish
More About this Item
These glass screw thread vials are designed specifically for autosamplers that require vials to hang from trays or supports. The wide neck of the vial, and the wider cap profile ensure that vials hang securely.
Vials with write-on patches have graduations at 0.5, 1.0, and 1.5 mL. Caps have a 10-425 thread finish.