Target DP convenience kit with clear vials 98810-00 and caps 98810-21

Representative image only.
Target Mfr # C5000-80 – Item # EW-98810-90
Save time during sample preparation and reduce the risk of contamination
  • Vials are constructed of clear glass

PRODUCT OPTIONS

$86.00USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)2
  • Tube Bottom ShapeFlat
  • Cap TypeScrew-Thread
  • Cap MaterialIvory PTFE
  • ThreadExternal
  • Septum TypeRed rubber
  • Height (in)1 17/64
  • Outside Diameter (in)15/32
  • Height (mm)32
  • Outside Diameter (mm)12
  • DescriptionDP convenience kit with clear vials 98810-00 and caps 98810-21

Key Features

  • Vials are constructed of clear glass

More About this Item

These easy-order, pre-selected kits include vials and caps with septa. Convenience kits include 100 of each component bulk-packed in storage compartments—keeping vials and caps handy and free of dust and contamination.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat