Taylor 1443 Pro Series Refrigerator / Freezer Digital Thermometer
Representative image only.
Taylor Mfr # 1443 – Item # EW-90001-31
Help avoid food spoilage with a thermometer in your refrigerator and freezer.
  • Temperature range -4 to 140°F (20 to 60°C)
  • Bold 0.8” digits are easy to read.
  • Hangs, stands or attaches with magnet.
  • 1 year warranty
$6.48USD / Each
$8.10USD / Each
$88.50 - $161.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-4
  • Max Temperature (° F)140
  • Min Temperature (° C)-20
  • Max Temperature (° C)60
  • Accuracy2%
  • Memory CapacityMin/Max
  • Mounting TypeClip, stand, or magnet
  • Display TypeDigital
  • Battery1 CR2023 (included)
  • DescriptionRefrigerator / Freezer Digital Thermometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Temperature range -4 to 140°F (20 to 60°C)
  • Bold 0.8” digits are easy to read.
  • Hangs, stands or attaches with magnet.
  • 1 year warranty
More About this Item
Know the ambient temperature of your refrigeration units with a glance with the Taylor 1443 digital refrigerator / freezer thermometer! This compact thermometer displays temperature from -4 to 140 degrees Fahrenheit on a large 1" display so you can view it from a distance. It can conveniently switch between Fahrenheit and Celsius measurements, and back-mounted magnets let you pin this thermometer on a freezer's interior wall. It also comes with a foldable hanger so that it can attach to the bottom of a wire shelf or stand on its own.