Taylor 3510 TruTemp Series Candy / Deep Fry Analog glass Tube Thermometer with Immersion Shield
Representative image only.
Taylor Mfr # 3510 – Item # EW-90001-34
Use this thermometer to ensure a consistent final product every time!
  • Adjust immersion depth with the included clip
  • Tempered glass tube.
  • Temperature range 100 to 400°F (50 to 200°C).
  • Insulated pan clip and safety cap.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)100
  • Max Temperature (° F)400
  • Min Temperature (° C)50
  • Max Temperature (° C)200
  • Accuracy2%
  • DescriptionCandy / Deep Fry Analog glass Tube Thermometer with Immersion Shield
  • Warranty1 year
1 Year Warranty
Key Features
  • Adjust immersion depth with the included clip
  • Tempered glass tube.
  • Temperature range 100 to 400°F (50 to 200°C).
  • Insulated pan clip and safety cap.
  • 1 year warranty
More About this Item
An essential addition to commercial kitchens, this thermometer allows you to easily check the temperatures of delicate candy mixes or fryer oils so your final food products come out just right.
The 3510 TruTemp Candy/Deep Fry Thermometer is an easy-to-use thermometer with an appealing design. Use this kitchen thermometer to get a good reading on your candy mixture or to monitor deep-fry oil temperature as you cook.
The tempered glass tube design of the 3510 TruTemp Candy/Deep Fry Thermometer allows the thermometer to extend into your pot and gives you a temperature reading in Fahrenheit from 100 to 400 degrees, and in Celsius, from 50 to 200 degrees. These readings are not labeled with heat stages for candy making, so you will need to have a goal temperature in mind and keep a sharp eye on your thermometer as you cook.