Taylor 501 Connoisseur Gourmet Bimetal Test Thermometer, 0/220°F
Representative image only.
Taylor Mfr # 501 – Item # EW-90001-21
Case additive inhibits bacterial grpwth
  • Ideal for food applications
  • NSF-listed
  • Multi-purpose protective case also functions as a handle and recalibration wrench
  • Bold easy-to-read dial numerals
  • Temperature ranges from 20 to 220°F.
  • 1-7/8" thermometer head with a 1-1/4" viewing dial.
  • Stainless steel construction with silicone grips for safe and easy handling.
  • Innovative shape and design ensures the thermometer will not roll off counters.
  • Includes a stem case with a convenient pocket clip.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)20
  • Max Temperature (° F)220
  • Dial Size (in)1.25
  • Stem Length (in)6
  • DescriptionGourmet Bimetal Test Thermometer, 0/220°F
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for food applications
  • NSF-listed
  • Multi-purpose protective case also functions as a handle and recalibration wrench
  • Bold easy-to-read dial numerals
  • Temperature ranges from 20 to 220°F.
  • 1-7/8" thermometer head with a 1-1/4" viewing dial.
  • Stainless steel construction with silicone grips for safe and easy handling.
  • Innovative shape and design ensures the thermometer will not roll off counters.
  • Includes a stem case with a convenient pocket clip.
  • 1 year warranty
More About this Item
nstant read, multi-purpose thermometer. Use for various cooking activites to ensure safe and healthy meals.