This item is now discontinued. We recommend the following as suggested replacements: EW-90001-41
Taylor 519 Connoisseur Series Gourmet Deep Fry / Candy Digital Thermometer
Representative image only.
Taylor Mfr # 51966 – Item # EW-90001-42
Perfect your frying and candy-making recipes
  • Presets for 11 different candy and deep fry temperatures.
  • Unit beeps and flashes when desired preset temperature is approaching and then reached.
  • Head of thermometer can be tilted for easy reading from any angle or height.
  • Pan clip to hold thermometer to a cooking pot.
  • Silicone grips make it easy to remove from hot pan or deep fryer.
  • Includes a silicone clip to clip to the side of a pan.
  • Stem 5.5" long.
  • Auto shut off to conserve batteries.
  • Temperatures range -40 to 450F (-40 to 230C).
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-40
  • Max Temperature (° F)450
  • Min Temperature (° C)-40
  • Max Temperature (° C)230
  • Accuracy2%
  • AlarmYes
  • Display TypeDigital
  • Diameter (in)1 3/4
  • Stem Length (in)5
  • DescriptionGourmet Deep Fry / Candy Digital Thermometer
  • Warranty1 year
1 Year Warranty
Includes
Lithium battery
Key Features
  • Presets for 11 different candy and deep fry temperatures.
  • Unit beeps and flashes when desired preset temperature is approaching and then reached.
  • Head of thermometer can be tilted for easy reading from any angle or height.
  • Pan clip to hold thermometer to a cooking pot.
  • Silicone grips make it easy to remove from hot pan or deep fryer.
  • Includes a silicone clip to clip to the side of a pan.
  • Stem 5.5" long.
  • Auto shut off to conserve batteries.
  • Temperatures range -40 to 450F (-40 to 230C).
  • 1 year warranty
More About this Item
Connoisseur Digital Deep-Fry/Candy Thermometer has a 6" (15.2 cm) stem with a silicone-sheathed pan clip to protect cookware. This thermometer is programmed with eleven presets for common deep fry and candy temperatures. When set point temperature is reached, the thermometer beeps and flashes. Adjustable head is viewable from any angle and features silicone grips to make handling easy when you have wet hands or gloves. Dual-line 1-3/4" (4.5 cm) LCD shows current and set point temperatures simultaneously.