Taylor 6092NFSA Commercial Series Color Coded Cross-Contamination Bimetal Pocket Thermometers, Set of Five
Representative image only.
Taylor Mfr # 6092NFSA – Item # EW-90001-53
Color Coded Thermometers Food Themometers
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
  • Thermometer Colors:
  • - Brown (cooked meats)
  • - Blue (fish)
  • - Purple (allergens)
  • - Red (raw meat)
  • - Yellow (poultry)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)0
  • Max Temperature (° F)200
  • Stem Length (in)5
  • DescriptionColor Coded Cross-Contamination Bimetal Pocket Thermometers, Set of Five
  • Warranty1 year
1 Year Warranty
Key Features
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
  • Thermometer Colors:
  • - Brown (cooked meats)
  • - Blue (fish)
  • - Purple (allergens)
  • - Red (raw meat)
  • - Yellow (poultry)
  • 1 year warranty
More About this Item
Color coded food safety pocket thermometer. This Taylor Precision 6092N, also known as a temperature indicator, is constructed of stainless steel. It has a 1" diameter dial and a 5" stem. This shatter-proof Taylor Precision 6092N has a pocketed sleeve. The sleeve features a unique built-calibration feature. This thermometer, Taylor Precision 6092N reads temperatures ranging from 0°F to +220°F. It is NSF listed. This Taylor Precision 6092N is available in five colors. Kitchens can use these pocket thermometers to color code. This food safe Taylor Precision 6092N helps to prevent cross-contamination. Purple is ideal for allergens, yellow for poultry, red for raw meat, brown for cooked meat and blue for fish.
Ensure the food you're serving is always at a safe and proper temperature with the Taylor 6092NFSA 5-pack of thermometers, which comes with five available colors to prevent cross-contamination across a variety of meats! The extremely accurate components can measure temperatures from 0-220 degrees Fahrenheit, within two degrees of accuracy. These thermometers also feature the exclusive Safe-T-Set recalibration method and calibration wrench to ensure you're always getting an accurate reading!
The 5-pack includes various colored thermometers to designate kitchen usage, and will help ensure employees don't cross-contaminate the thermometer with other meats and food products during the cooking process. The 5" stainless steel stem and shatter-proof polycarbonate crystal lens will allow you to use this dependable thermometer for years to come!