Taylor 6092NPR Color Coded Cross-Contamination Bimetal Pocket Thermometer - Purple, Food Allergy
Representative image only.
Taylor Mfr # 6092NPRBC – Item # EW-90025-74
Color Coded Thermometers Food Themometers
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)0
  • Min Temperature (° C)-10
  • Max Temperature (° F)220
  • Max Temperature (° C)100
  • Division (° C)2
  • Division (° F)2
  • DescriptionColor Coded Cross-Contamination Bimetal Pocket Thermaometer - Purple, Food Allergy
  • Warranty1 year
1 Year Warranty
Key Features
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
  • 1 year warranty
More About this Item
Color coded food safety pocket thermometer. This Taylor Precision 6092N, also known as a temperature indicator, is constructed of stainless steel. It has a 1" diameter dial and a 5" stem. This shatter-proof Taylor Precision 6092N has a pocketed sleeve. The sleeve features a unique built-calibration feature. This thermometer, Taylor Precision 6092N reads temperatures ranging from 0°F to +220°F. It is NSF listed. This Taylor Precision 6092N is available in five colors. Kitchens can use these pocket thermometers to color code. This food safe Taylor Precision 6092N helps to prevent cross-contamination. Purple is ideal for allergens, yellow for poultry, red for raw meat, brown for cooked meat and blue for fish.