TBS MP-B Black marking pen for labeling slides or cassettes; 12/pk
Representative image only.
TBS Mfr # MP-B – Item # EW-48579-70
The laboratory marking pen with priming action and replaceable felt tip
  • Quick drying
  • Reagent resistant
  • Replaceable tips
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Tip StyleFine-point
  • Pen ColorBlack
  • DescriptionBlack marking pen for labeling slides or cassettes; 12/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Quick drying
  • Reagent resistant
  • Replaceable tips
  • 1 year warranty
More About this Item
Use these marking pens to labeling slides or cassettes. Quick drying ink will not smudge. Reistant to even the toughest reagents, including alchohol and xylene. Once your pen wears out simply replace the new tip. Choose from red or black.