Techne DNA Lactobacillus plantarum, without mastermix
Representative image only.
Techne Item # EW-93959-59
Designed for the in vitro quantification of human gastrointestinal microbiome
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedNo
  • DescriptionLactobacillus plantarum, without mastermix
Includes
Lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Each kit is specifically designed to ensure the broadest possible detection profile and to detect all relevant strains and subtypes. All kits are available with onestep mastermix or without.