Cole-Parmer Dri-Block® Aluminum Heating Block Insert, 96-well plate, round bottom
Representative image only.
Cole-Parmer Item # EW-36620-69
Interchangeable heating blocks offer excellent thermal conductivity
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number Of Blocks1
  • Block Number Of Openings96
  • Block TypeAluminum
  • DescriptionDri-Block® Aluminum Heating Block Insert, 96-well plate, round bottom
  • Warranty1 Year
1 Year Warranty
Key Features
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.