Cole-Parmer F840D Polypropylene Bath Balls, 25 mm Diameter; 250/PK
Representative image only.
Cole-Parmer Item # EW-12122-76
Enhance the performance of your Cole-Parmer® baths and circulators
  • Constructed of durable polypropylene
  • 1 year warranty
  • Previous part number F840D
PRODUCT OPTIONS
$114.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Compatible WithAll water baths
  • Diameter (mm)25
  • DescriptionPolypropylene Bath Balls, 25 mm Diameter; 250/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable polypropylene
  • 1 year warranty
  • Previous part number F840D
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
A ball blanket is an effective way of reducing evaporation and loss of heat from a water bath. It acts as effectively as a lid, whilst providing instant access to the bath. The 25mm diameter spheres are supplied in packs of 250.