Cole-Parmer FGABLE08 Bath Lid, Gabled, for 8 Liter Baths
Representative image only.
Cole-Parmer Item # EW-12122-95
Enhance the performance of your Cole-Parmer® baths and circulators
  • 3 year warranty
  • Previous part number FGABLE08
PRODUCT OPTIONS
$437.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithUnheated water bath 16104-53
  • DescriptionBath Lid, Gabled, for 8 Liter Baths
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
  • Previous part number FGABLE08
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Manufactured from stainless steel and available to fit 8 liter baths to help prevent evaporation losses. Gabled lids provide extra working headroom within the bath.