Cole-Parmer FGABLE12 Bath Lid, Gabled, for 12 Liter Baths
Representative image only.
Cole-Parmer Item # EW-12122-96
Enhance the performance of your Cole-Parmer® baths and circulators
  • 3 year warranty
  • Previous part number FGABLE12
PRODUCT OPTIONS
$536.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithUnheated water bath 16101-88
  • DescriptionBath Lid, Gabled, for 12 Liter Baths
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
  • Previous part number FGABLE12
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Manufactured from stainless steel and available to fit 12 liter baths to help prevent evaporation losses. Gabled lids provide extra working headroom within the bath.