Tedia Diethylene Glycol, Certified Grade; 6 x 1 L, Amber Glass

Representative image only.
Tedia Mfr # DR0429-002 – Item # EW-15000-49
High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)6000
  • PackagingAmber Glass
  • CAS Number111-46-6
  • Shelf Life (months)60
  • DescriptionDiethylene Glycol, Certified Grade; 6 x 1 L, Amber Glass

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat