Tedia Glacial Acetic Acid, ACS Reagent Grade; 20 L, Poly Pail

Representative image only.
Tedia Mfr # AR0104-031 – Item # EW-15000-11
High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.

PRODUCT OPTIONS

$346.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • FormLiquid
  • Volume (mL)20000
  • PackagingPoly Pail
  • CAS Number64-19-7
  • UN NumberUN2789
  • Shelf Life (months)36
  • DescriptionGlacial Acetic Acid, ACS Reagent Grade; 20 L, Poly Pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat