Tedia Heptanesulfonic Acid, Sodium Salt, HPLC/Spectrophotometric Grade, 25g

Representative image only.
Tedia Mfr # HS3403-009 – Item # EW-15001-03
High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC and Spectrophotometric
  • ChemistryAcids
  • FormSolid
  • Weight (g)25
  • Formula Weight202.25
  • PackagingAmber Glass Bottle
  • CAS Number22767-50-6
  • Shelf Life (months)36
  • DescriptionHeptanesulfonic Acid, Sodium Salt, HPLC/Spectrophotometric Grade, 25g

Key Features

  • Quality and consistency your lab can count on

More About this Item

1-Heptanesulfonic acid sodium salt is a white crystalline or flaky solid used as a surfactant chromatography conditioner or lab reagent in biological studies.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat