Tedia MS1922-006 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC SKU 1500135

Representative image only.
Tedia Mfr # MS1922-006 – Item # EW-15001-35
High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC and Spectrophotometric
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)16000
  • PackagingPVC
  • UN NumberUN1230
  • CAS Number67-56-1
  • Shelf Life (months)60
  • DescriptionMethyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat