This item is now discontinued. View the direct replacement: EW-21940-43 or browse similar items
Diba Cones, 1/16", 10 per pack
Representative image only.
Diba Mfr # 008CZ16 – Item # EW-06471-56
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPTFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • DescriptionCones, 1/16\", 10 per pack
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use these fittings with tubing ranging from 1 to 11 mm OD. All standard fittings except models 06473-07 and -08 come with polypropylene (PP) caps, two large VITON O-rings per cap to connect 2 to 4 mm OD tubing, one small VITON O-ring per cap (use together with large O-rings) to connect 1 to 2 mm OD tubing, and one PTFE cone per cap to make chemically inert connections for 1/16" OD tubing or to create zero dead volume fittings for connection to high-pressure fittings (sold below).
Standard fittings 06473-07 and -08 include one fluoroelastomer O-ring per PTFE cap. All chemically inert fittings come with TEFZEL® caps and 1/16" tube OD PTFE cones for improved chemical resistance. All O-rings are made of fluoroelastomer. Max fluid pressure (up to 50 psi) will vary depending on tubing material and size.