Testo 0632 1535 Replacement IAQ Probe 0 to +50 °C
Representative image only.
Testo Mfr # 0632 1535 – Item # EW-37486-10
Optimize performance of your air/gas analyzer
  • Replacement probes
  • Data logging software
  • 4 functions in one probe — measurement of CO2, humidity, temperature and absolute pressure
  • Rapid checking of the indoor climate
  • Prevention of poor air quality in indoor rooms via CO2 measurement
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMultifunction air measurement meter 37489-01
  • ConnectionsMiniDIN 
  • DescriptionReplacement IAQ Probe 0 to 50 °C
Two year warrantyConformite Europeenne
Key Features
  • Replacement probes
  • Data logging software
  • 4 functions in one probe — measurement of CO2, humidity, temperature and absolute pressure
  • Rapid checking of the indoor climate
  • Prevention of poor air quality in indoor rooms via CO2 measurement
  • 2 year warranty
More About this Item
Replacement probes, software, and other accessories to customize your sampling and analysis system.
Designed to be used in applications that require multiple measurements, the IAQ simultaneously monitors CO2, relative humidity and temperature. The IAQ Probe has a MiniDIN connection. This probe features a temperature range of 32.0° to 122.0 °F (0 to +50 °C) with an accuracy of ±0.5 °F (±0.3 °C). The humidity range is 0 to 100% RH with an accuracy of ±2 %RH (+2 to +98 %RH). The absolute pressure range is +240 to +460 InH₂O with an accuracy of ±4 InH₂O. The Ambient CO₂ range is 0 to 10,000 ppm. Multifunction Air Measurement Meter model 37486-01 sold separately.