Testo 0632 3173 Ambient CO Stick, 0 to 1999 ppm
Representative image only.
Testo Mfr # 0632 3173 – Item # EW-90449-79
Compact CO meter with self-test function
  • No zeroing rquired, just turn it on to measure
  • Three year warranty on CO sensor
  • Visual and audible alarm
  • Adjustable alarm settings
  • Self test mode verifies CO sensor anytime
$275.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredCO
  • Min CO Range (ppm)0
  • Max CO Range (ppm)1999
  • DescriptionAmbient CO Stick, 0 to 1999 ppm
Key Features
  • No zeroing rquired, just turn it on to measure
  • Three year warranty on CO sensor
  • Visual and audible alarm
  • Adjustable alarm settings
  • Self test mode verifies CO sensor anytime
More About this Item
This ambient CO Meter with user defined audible alarms makes this pocket size meter perfect for personal safety for identifying potentional CO sources. With this instrument, the user can perform a weekly sensor test to assure for accurate readings without the use of test gas.