Testo 805i Smart Probe Infrared Thermometer for use with Smartphone or Tablet
Representative image only.
Testo Mfr # 0560 1805 01 – Item # EW-30009-43
View HVAC measurement on your Smart device from up to 60 feet away!
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • Measurement area indicated by circular laser pattern
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-22
  • Max Temperature (° F)482
  • Min Temperature (° C)-30
  • Max Temperature (° C)250
  • Accuracy±2.7 °F or ±1.5 % of reading (32 to 482 °F), ±3.6 °F (-4 to 32 °F), ±4.5 °F (-22 to -4 °F)
  • Resolution0.1°
  • Low Battery IndicatorYes
  • BatteryThree AAA batteries (included)
  • DescriptionSmart Probe Infrared Thermometer for use with Smartphone or Tablet
Two year warrantyApple App StoreConformite EuropeenneGoogle Play App
Includes
Three AAA batteries and certificate of calibration.
Key Features
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • Measurement area indicated by circular laser pattern
  • 2 year warranty
More About this Item
Ths compact, rugged, easy-to-use wireless infrared thermometer is ideal for noncontact temperature readings of breakers, motors, ducts, and registers – ideal for the HVAC professional. It features a 10:1 optic target ratio and uses a circular laser pattern to accurately measure a specific area.
Easily monitor and transmit all measured data wirelessly via Bluetooth® to your smartphone or tablet from up to 60 feet away. With the free Testo Smart Probes app, you can display measurements from up to six Smart Probes at the same time, visualize measurement value changes as a graph or a table, and perform automatic calculations. Easily turn your data into a custom report, attach images created with the app, and send via email as a PDF or Excel® file.
Combine this probe with the 405i Smart Probe hotwire anemometer, 410i Smart Probe vane anemometer, and 605i Smart Probe thermohygrometer to monitor HVAC-R applications.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
However, none of our infrared thermometers, no matter the manufacture, are approved for medical usage, and are not FDA approved.
For help in selecting an infrared thermometer for industrial, food or laboratory usage please contact the customer service team at Cole-Parmer.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.