This item is now discontinued. Please browse similar items
Texas Electronics 2-200 Analog Wind Speed System, 0 to 100 mph, self-powered
Representative image only.
Texas Electronics Mfr # 2-200 – Item # EW-99535-71
SPECS & DESCRIPTION
Specifications & Description
  • Max Wind Speed (mph)100
  • Min Wind Speed (mph)0
  • DescriptionAnalog Wind Speed System, 0 to 100 mph, self-powered
More About this Item
The Model 2-200 Wind Indicating System is an extremely accurate, low cost wind measuring system. This system consists of an anemometer (Model TV-114) and a dial indicator. This system is self-generating; hence no external power source or battery is required. The indicator is contained in a cast aluminum housing with universal mounting brackets. These brackets greatly simplify meter mounting in marine and mobile installations as they provide for top, back, or bottom mounting. The TV-114 anemometer is a rugged sensor constructed of gold anodized aluminum exterior, contains a stainless steel lubricated bearings, and a brushless tachometer type AC generator. The indicator converts and scales to one of the following ranges: 0 to 100 MPH, 0 to 100 knots, 0 to 160 km/hr, and 0 to 50 meters/sec The Model 2-200 comes standard with 60 feet of cable. However, cable can be increased to one thousand feet without noticeable impact on performance. Indicator Size: 4.5" wide, 3.5" high, 2" deep Sensor Size: 7" high, 18" cup wheel diameter
FEATURES
  • Self-generating system requires no external power source
  • Provides for safe operation of wind-affected equipment
  • Rugged instruments can withstand extreme conditions
  • Extremely accurate readings at economical price
  • Mounts easily in a variety of configurations
  • Available in a variety of ranges
  • Over 30 years in production