Thermo Scientific 222056 Bottle Gripper, for large and medium 35mm bottles
Representative image only.
Thermo Scientific Mfr # 222056 – Item # EW-13247-37
Manufactured using thick walled borosilicate glass to ensure safe and easy handling
  • Improves the flow of probe solution around and through membranes for optimal results
  • Designed to exacting safety standards, these bottles have screw caps containing flat PTFE seals to ensure leakresistant hybridization
  • Full range of bottles accommodates every size of membrane
  • Enables multiple blots to be processed simultaneously in one 35mm diameter bottle without direct overlap
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBottle Gripper, for large and medium 35mm bottles
  • Warranty1 year
1 Year Warranty
Key Features
  • Improves the flow of probe solution around and through membranes for optimal results
  • Designed to exacting safety standards, these bottles have screw caps containing flat PTFE seals to ensure leakresistant hybridization
  • Full range of bottles accommodates every size of membrane
  • Enables multiple blots to be processed simultaneously in one 35mm diameter bottle without direct overlap
  • 1 year warranty
More About this Item
Choose from bottles, mesh, and bottle grippers to complete your hybridization and mesh system. Bottle grippers are ideal for safe and easy handling of bottles.
Bottle gripper ensures safe and easy handling of hybridization bottles (sold separately).