Thermo Scientific 3551-1 Explosion Proof Refrigerator/Freezer; Two Doors; 230V

Representative image only.
Thermo Scientific Mfr # 10ECEETSV – Item # EW-44200-05
Combination storage for biological materials or chemicals
  • CFC-free insulation and coolant
  • 15 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)13.4
  • Freezer Capacity (Liters)379
  • Min Temperature - Freezer (° F)-40
  • Max Temperature - Freezer (° F)14
  • Min Temperature - Freezer (° C)-40
  • Max Temperature - Freezer (° C)-10
  • Number of Shelves4
  • Power (VAC)230
  • Power (Hz)50
  • Power (Breaker)15 A
  • DescriptionExplosion Proof Refrigerator/Freezer; Two Doors; 240V
  • Warranty15 month
15 Month WarrantyCanadian Standards AssociationUL Listed.

Key Features

  • CFC-free insulation and coolant
  • 15 month warranty

More About this Item

Explosion-Proof Models protect against explosions both inside and outside of the unit. All motors, switches, and thermostats prohibit arcing that can ignite flammable air-vapor mixtures. They meet Class 1, Group C and D requirements for hazardous environments. Explosion-proof units must be hard-wired to voltage source.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat