Thermo Scientific 3551-10 Flammable Materials Refrigerator/Freezer, Two-Door; 115V

Representative image only.
Thermo Scientific Mfr # 10FCEETSA – Item # EW-44201-00
Combination storage for biological materials or chemicals
  • CFC-free insulation and coolant
  • 15 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)24.4
  • Freezer Capacity (Liters)691
  • Min Temperature - Freezer (° F)-40
  • Max Temperature - Freezer (° F)14
  • Min Temperature - Freezer (° C)-40
  • Max Temperature - Freezer (° C)-10
  • Number of Shelves4
  • Power (VAC)115
  • Power (Hz)50
  • Power (Breaker)15 A
  • DescriptionFlammable Materials Refrigerator/Freezer, Two-Door; 120V
  • Warranty15 month
15 Month WarrantyCanadian Standards Association

Key Features

  • CFC-free insulation and coolant
  • 15 month warranty

More About this Item

Flammable Materials Models contain no internal electrical devices that can trigger the explosion of hazardous materials. They are supplied with a 6-ft (1.8-m) cord with three-prong plug.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat