Thermo Scientific 403Q 5.1 cu ft. Gravity-Convection Incubator - 120V
Representative image only.
Thermo Scientific Mfr # 403Q – Item # EW-03610-04
High quality incubators at affordable prices
  • Adjustable hydraulic thermostat reacts quickly to temperature changes
  • Radiant warm wall heating provides maximum temperature uniformity in chamber
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (cu ft)5.1
  • Capacity (Liters)144
  • Power (VAC)120
  • Power (Hz)50/60
  • Description5.1 cu ft. Gravity-Convection Incubator - 120V
  • Warranty2 year
Two year warrantyCanadian Standards AssociationUL Listed.
Key Features
  • Adjustable hydraulic thermostat reacts quickly to temperature changes
  • Radiant warm wall heating provides maximum temperature uniformity in chamber