Thermo Scientific 66002-001 GLD Pro Gas Leak Detector
Representative image only.
Thermo Scientific Mfr # 66002-001 – Item # EW-81949-10
Check for minute leaks in gas chromatography systems
  • Zero the unit by simply pressing a button in a fresh air environment
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredGeneral Purpose
  • Carbon Dioxide Sensitivity1.0 x 10-4 cc/sec
  • Argon Sensitivity1.0 x 10-4 cc/sec
  • Hydrogen Sensitivity1.0 x 10-5 cc/sec
  • Helium Sensitivity1.0 x 10-5 cc/sec
  • Nitrogen Sensitivity1.4 x 10 ^-3 cc/sec
  • Battery TypeRechargeable batteries (included)
  • Battery Life6 hours typical
  • DescriptionGLD Pro Gas Leak Detector
  • Warranty1 year
1 Year Warranty
Key Features
  • Zero the unit by simply pressing a button in a fresh air environment
  • 1 year warranty
More About this Item
The Thermo Scientific GLD Pro leak detector detects leaks of any gas with a thermal conductivity different from air. Leaks are indicated by LED indicators; when the last indicator lights, an audible alarm will sound. Use the optional small probe adapter for hard-to-reach leaks.