Thermo Scientific 75003095 Centrifuge Tube Adapter; 100 mL to 15 mL Falcon Tube; Each
Representative image only.
Thermo Scientific Mfr # 75003095 – Item # EW-17702-84
$131.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Tube Sizes Accepted15 to 100 mL falcon tube
  • Number Of Tubes Accepted1
  • DescriptionCentrifuge Tube Adapter; 100 mL to 15 mL Falcon Tube; Each
More About this Item
Keep your centrifuge in good working order with these accessories.
Compatible with the HIGHConic rotor, HIGHConic II rotor, and Fiberlite F15-6x100y rotor.